missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C9orf39 Partial ORF (NP_060208, 971 a.a. - 1070 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_060208
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 54875
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16137776
View Documents Promotion Details
Abnova™
H00054875-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16127776
View Documents Promotion Details
Abnova™
H00054875-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

Sequence: QNDVHVVRRQIRELKKMKKNRDACKTSTHKAQTLAASILNISRSDLEEILDTEDQVEIEKTKIDAENDKEWMLYIQKLLEGQSLALSPRLKCNGAIMAHQ
Specifications

Specifications

NP_060208
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C9orf101/C9orf39/FLJ20276/FLJ25636/RP11-340N12.1/bA340N12.1
CNTLN
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
54875
C9orf39 (Human) Recombinant Protein (Q01)
QNDVHVVRRQIRELKKMKKNRDACKTSTHKAQTLAASILNISRSDLEEILDTEDQVEIEKTKIDAENDKEWMLYIQKLLEGQSLALSPRLKCNGAIMAHQ
RUO
CNTLN
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications