missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C5orf4 Full-length ORF (NP_115761.1, 1 a.a. - 191 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010826-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195742

  • 349.00€ / 10µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

Sequence: MKGEAGHMLHNEKSKQEGHIWGSMRRTAFILGSGLLSFVAFWNSVTWHLQRFWGASGYFWQAQWERLLTTFEGKEWILFFIGAIQVPCLFFWSFNGLLLVVDTTGKPNFISRYRIQVGKNEPVDPVKLRQSIRTVLFNQCMISFPMVVFLYPFLKWWRDPCRRELPTFHWFLLELAIFTLIEEVLFYYSHR
Specifications

Specifications

NP_115761.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ13758
C5orf4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10826
C5orf4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKGEAGHMLHNEKSKQEGHIWGSMRRTAFILGSGLLSFVAFWNSVTWHLQRFWGASGYFWQAQWERLLTTFEGKEWILFFIGAIQVPCLFFWSFNGLLLVVDTTGKPNFISRYRIQVGKNEPVDPVKLRQSIRTVLFNQCMISFPMVVFLYPFLKWWRDPCRRELPTFHWFLLELAIFTLIEEVLFYYSHR
RUO
C5orf4
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers