missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C17orf57 Full-length ORF (AAH37876.1, 1 a.a. - 531 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number AAH37876.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 124989
Molecular Weight (g/mol) 86.2kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16117663
View Documents Promotion Details
Abnova™
H00124989-P01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16107663
View Documents Promotion Details
Abnova™
H00124989-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

Sequence: MENDDLESKRPKNTWQIRKFLGGVGSSNVGVQEPYSKNGINFKKHSEKGEIHDSKSKPQSLKSSTSLSKSLDKSDISSIPKLQKPAVRKHSSLQKQVSSTEKTAISTLENFCEAISKLQENYIAAEELQSILPSTGINLLDEEFQKIVTDTSRNENGMVELDDFVNALAKERSFPECNALPGVIKAIDKIKDKNVDYEDLNTCLQNFGIYLSKPEFKKITELTEAGETKKVNFKEFIDTMMSNTECFSEKLVLPDAIETLDDLRKETMSVSDLWNTLSSLNSNLKKDEFLAALELVTVDEGDKVQFEEFAKVVRNMRDAARLEELQEVVLAADLLEGDMIAGKNLEDFLRNVGIKSPKEEVEKILQSDFVSEDNMVNIKDCMRALRDTQKFSNYIDFRKEASNLKLPKVNEIKEAANILSHVDNGKIGIPDLEHALKCLNVNLTEEDFNEALNCCNVSDNMEVDLKDFLMKMKESPHFQKSKATQILLATTQILQNDLVDVSDLKTLLMDKDLHTANAILTVMLRHVPEHG
Specifications

Specifications

AAH37876.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
86.2kDa
Glutathione Sepharose 4 Fast Flow
MENDDLESKRPKNTWQIRKFLGGVGSSNVGVQEPYSKNGINFKKHSEKGEIHDSKSKPQSLKSSTSLSKSLDKSDISSIPKLQKPAVRKHSSLQKQVSSTEKTAISTLENFCEAISKLQENYIAAEELQSILPSTGINLLDEEFQKIVTDTSRNENGMVELDDFVNALAKERSFPECNALPGVIKAIDKIKDKNVDYEDLNTCLQNFGIYLSKPEFKKITELTEAGETKKVNFKEFIDTMMSNTECFSEKLVLPDAIETLDDLRKETMSVSDLWNTLSSLNSNLKKDEFLAALELVTVDEGDKVQFEEFAKVVRNMRDAARLEELQEVVLAADLLEGDMIAGKNLEDFLRNVGIKSPKEEVEKILQSDFVSEDNMVNIKDCMRALRDTQKFSNYIDFRKEASNLKLPKVNEIKEAANILSHVDNGKIGIPDLEHALKCLNVNLTEEDFNEALNCCNVSDNMEVDLKDFLMKMKESPHFQKSKATQILLATTQILQNDLVDVSDLKTLLMDKDLHTANAILTVMLRHVPEHG
RUO
C17orf57
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
124989
C17orf57 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ40342
C17orf57
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications