missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C14orf122 Full-length ORF (NP_057133.2, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_057133.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51016
Molecular Weight (g/mol) 49.5kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16119512
View Documents Promotion Details
Abnova™
H00051016-P01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16109512
View Documents Promotion Details
Abnova™
H00051016-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

Sequence: MGEVEISALAYVKMCLHAARYPHAAVNGLFLAPAPRSGECLCLTDCVPLFHSHLALSVMLEVALNQVDVWGAQAGLVVAGYYHANAAVNDQSPGPLALKIAGRIAEFFPDAVLIMLDNQKLVPQPRVPPVIVLENQGLRWVPKDKNLVMWRDWEESRQMVGALLEDRAHQHLVDFDCHLDDIRQDWTNQRLNTQITQWVGPTNGNGNA
Specifications

Specifications

NP_057133.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.5kDa
Glutathione Sepharose 4 Fast Flow
MGEVEISALAYVKMCLHAARYPHAAVNGLFLAPAPRSGECLCLTDCVPLFHSHLALSVMLEVALNQVDVWGAQAGLVVAGYYHANAAVNDQSPGPLALKIAGRIAEFFPDAVLIMLDNQKLVPQPRVPPVIVLENQGLRWVPKDKNLVMWRDWEESRQMVGALLEDRAHQHLVDFDCHLDDIRQDWTNQRLNTQITQWVGPTNGNGNA
RUO
FAM158A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
51016
C14orf122 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C14orf122/CGI-112
FAM158A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications