missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BCAM Partial ORF (NP_005572, 32 a.a. - 131 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_005572 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4059 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127481
|
Abnova™
H00004059-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16117481
|
Abnova™
H00004059-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: EVRLSVPPLVEVMRGKSVILDCTPTGTHDHYMLEWFLTDRSGARPRLASAEMQGSELQVTMHDTRGRSPPYQLDSQGRLVLAEAQVGDERDYVCVVRAGASpecifications
NP_005572 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AU/CD239/LU/MSK19 | |
BCAM | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4059 | |
BCAM (Human) Recombinant Protein (Q01) | |
EVRLSVPPLVEVMRGKSVILDCTPTGTHDHYMLEWFLTDRSGARPRLASAEMQGSELQVTMHDTRGRSPPYQLDSQGRLVLAEAQVGDERDYVCVVRAGA | |
RUO | |
BCAM | |
Wheat Germ (in vitro) | |
GST | |
Liquid |