missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BAT5 Full-length ORF (NP_066983.1, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007920-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192192

  • 349.00€ / 10µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity. [provided by RefSeq]

Sequence: MAKLLSCVLGPRLYKIYRERDSERAPASVPETPTAVTAPHSSSWDTYYQPRALEKHADSILALASVFWSISYYSSPFAFFYLYRKGYLSLSKVVPFSHYAGTLLLLLAGVACLRGIGRWTNPQYRQFITILEATHRNQSSENKRQLANYNFDFRSWPVDFHWEEPSSRKESRGGPSRRGVALLRPEPLHRGTADTLLNRVKKLPCQITSYLVAHTLGRRMLYPGSVYLLQKALMPVLLQGQARLVEECNGRRAKLLACDGNEIDTMFVDRRGTAEPQGQKLVICCEGNAGFYEVGCVSTPLEAGYSVLGWNHPGFAGSTGVPFPQNEANAMDVVVQFAIHRLGFQPQDIIIYAWSIGGFTATWAAMSYPDVSAMILDASFDDLVPLALKVMPDSWRGLVTRTVRQHLNLNNAEQLCRYQGPVLLIRRTKDEIITTTVPEDIMSNRGNDLLLKLLQHRYPRVMAEEGLRVVRQWLEASSQLEEASIYSRWEVEEDWCLSVLRSYQAEHGPDFPWSVGEDMSADGRRQLALFLARKHLHNFEATHCTPLPAQNFQMPWHL
Specifications

Specifications

NP_066983.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
RUO
BAT5
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7920
BAT5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D6S82E/NG26
BAT5
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers