missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BAT5 Full-length ORF (NP_066983.1, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_066983.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7920 |
Molecular Weight (g/mol) | 89.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102202
|
Abnova™
H00007920-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16192192
|
Abnova™
H00007920-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity. [provided by RefSeq]
Sequence: MAKLLSCVLGPRLYKIYRERDSERAPASVPETPTAVTAPHSSSWDTYYQPRALEKHADSILALASVFWSISYYSSPFAFFYLYRKGYLSLSKVVPFSHYAGTLLLLLAGVACLRGIGRWTNPQYRQFITILEATHRNQSSENKRQLANYNFDFRSWPVDFHWEEPSSRKESRGGPSRRGVALLRPEPLHRGTADTLLNRVKKLPCQITSYLVAHTLGRRMLYPGSVYLLQKALMPVLLQGQARLVEECNGRRAKLLACDGNEIDTMFVDRRGTAEPQGQKLVICCEGNAGFYEVGCVSTPLEAGYSVLGWNHPGFAGSTGVPFPQNEANAMDVVVQFAIHRLGFQPQDIIIYAWSIGGFTATWAAMSYPDVSAMILDASFDDLVPLALKVMPDSWRGLVTRTVRQHLNLNNAEQLCRYQGPVLLIRRTKDEIITTTVPEDIMSNRGNDLLLKLLQHRYPRVMAEEGLRVVRQWLEASSQLEEASIYSRWEVEEDWCLSVLRSYQAEHGPDFPWSVGEDMSADGRRQLALFLARKHLHNFEATHCTPLPAQNFQMPWHLSpecifications
NP_066983.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
89.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
D6S82E/NG26 | |
BAT5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7920 | |
BAT5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
BAT5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |