missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human BACH1 Partial ORF (NP_996749, 396 a.a. - 492 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000571-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16013655

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a transcription factor that belongs to the cap'n'collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo- and/or hetero-oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. [provided by RefSeq]

Sequence: HLAKGFWSDICSTDTPCQMQLSPAVAKDGSEQISQKRSECPWLGIRISESPEPGQRTFTTLSSVNCPFISTLSTEGCSSNLEIGNDDYVSEPQQEPC
Specifications

Specifications

NP_996749
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HLAKGFWSDICSTDTPCQMQLSPAVAKDGSEQISQKRSECPWLGIRISESPEPGQRTFTTLSSVNCPFISTLSTEGCSSNLEIGNDDYVSEPQQEPC
RUO
BACH1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
571
BACH1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BACH1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers