missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AURKA Partial ORF (NP_940835, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00006790-Q01.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16024785

  • 341.70€ / 10µg

Estimated Shipment: 04-01-2022
Add to basket

Description

Description

The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MDRSKENCISGPVKATAPVGGPKRVLVTQQFPCQNPLPVNSGQAQRVLCPSNSSQRIPLQAQKLVSSHKPVQNQKQKQLQATSVPHPVSRPLNNTQKSKQPLPSAPENNP
Specifications

Specifications

NP_940835
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDRSKENCISGPVKATAPVGGPKRVLVTQQFPCQNPLPVNSGQAQRVLCPSNSSQRIPLQAQKLVSSHKPVQNQKQKQLQATSVPHPVSRPLNNTQKSKQPLPSAPENNP
RUO
AURKA
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
6790
AURKA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIK/ARK1/AURA/AURORA2/BTAK/MGC34538/STK15/STK6/STK7
AURKA
Yes
Videos
SDS
Documents
Certificates
Special Offers

Special Offers