missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ATPAF2 Full-length ORF (NP_663729.1, 1 a.a. - 289 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_663729.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 91647 |
Molecular Weight (g/mol) | 59.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16166633
|
Abnova™
H00091647-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16156633
|
Abnova™
H00091647-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 alpha subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. This gene is located within the Smith-Magenis syndrome region on chromosome 17. An alternatively spliced transcript variant has been described, but its biological validity has not been determined. [provided by RefSeq]
Sequence: MWRSCLRLRDGGRRLLNRPAGGPSASMSPGPTIPSPARAYAPPTERKRFYQNVSITQGEGGFEINLDHRKLKTPQAKLFTVPSEALAIAVATEWDSQQDTIKYYTMHLTTLCNTSLDNPTQRNKDQLIRAAVKFLDTDTICYRVEEPETLVELQRNEWDPIIEWAEKRYGVEISSSTSIMGPSIPAKTREVLVSHLASYNTWALQGIEFVAAQLKSMVLTLGLIDLRLTVEQAVLLSRLEEEYQIQKWGNIEWAHDYELQELRARTAAGTLFIHLCSESTTVKHKLLKESpecifications
NP_663729.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP12/ATP12p/LP3663/MGC29736 | |
ATPAF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
91647 | |
ATPAF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ATPAF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |