missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP7A Partial ORF (NP_000043, 1406 a.a. - 1500 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000538-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192081

  • 528.00€ / 25µg

Estimated Shipment: 10-11-2023
to see stock.
Add to basket

Description

Description

This gene encodes a transmembrane protein that functions in copper transport across membranes. The protein localizes to the trans-Golgi network, where it is predicted to supply copper to copper-dependent enzymes in the secretory pathway. The protein relocalizes to the plasma membrane under conditions of elevated extracellular copper and functions in the efflux of copper from cells. Mutations in this gene result in Menkes disease, X-linked cutis laxa, and occipital horn syndrome. [provided by RefSeq]

Sequence: FLKLYRKPTYESYELPARSQIGQKSPSEISVHVGIDDTSRNSPKLGLLDRIVNYSRASINSLLSDKRSLNSVVTSEPDKHSLLVGDFREDDDTAL
Specifications

Specifications

NP_000043
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.19kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FLKLYRKPTYESYELPARSQIGQKSPSEISVHVGIDDTSRNSPKLGLLDRIVNYSRASINSLLSDKRSLNSVVTSEPDKHSLLVGDFREDDDTAL
RUO
ATP7A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
538
ATP7A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ17790/MK/MNK
ATP7A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers