missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP6AP2 Full-length ORF (NP_005756.2, 1 a.a. - 350 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_005756.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10159
Molecular Weight (g/mol) 65.4kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16104582
View Documents Promotion Details
Abnova™
H00010159-P01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16194572
View Documents Promotion Details
Abnova™
H00010159-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases. [provided by RefSeq]

Sequence: MAVFVVLLALVAGVLGNEFSILKSPGSVVFRNGNWPIPGERIPDVAALSMGFSVKEDLSWPGLAVGNLFHRPRATVMVMVKGVNKLALPPGSVISYPLENAVPFSLDSVANSIHSLFSEETPVVLQLAPSEERVYMVGKANSVFEDLSVTLRQLRNRLFQENSVLSSLPLNSLSRNNEVDLLFLSELQVLHDISSLLSRHKHLAKDHSPDLYSLELAGLDEIGKRYGEDSEQFRDASKILVDALQKFADDMYSLYGGNAVVELVTVKSFDTSLIRKTRTILEAKQAKNPASPYNLAYKYNFEYSVVFNMVLWIMIALALAVIITSYNIWNMDPGYDSIIYRMTNQKIRMD
Specifications

Specifications

NP_005756.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.4kDa
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APT6M8-9/ATP6IP2/ATP6M8-9/ELDF10/HT028/M8-9/MGC99577/MRXE/MSTP009/XMRE
ATP6AP2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10159
ATP6AP2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
RUO
ATP6AP2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications