Learn More
Abnova™ Human ARPC4 Full-length ORF (NP_001020130.1, 1 a.a. - 78 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010093-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p20 subunit, has yet to be determined. Three transcript variants encoding two distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: MRFMMMRAENFFILRRKPVEGYDISFLITNFHTEQMYKHKLVDFVIHFMEEIDKEISEMKLSVNARARIVAEEFLKNFSpecifications
NP_001020130.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ARPC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10093 | |
ARPC4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARC20/MGC13544/p20-Arc | |
ARPC4 | |
Recombinant | |
wheat germ expression system |