missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARL6IP5 Partial ORF (NP_006398, 1 a.a. - 64 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_006398 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10550 |
Molecular Weight (g/mol) | 32.78kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102776
|
Abnova™
H00010550-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16192766
|
Abnova™
H00010550-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Expression of this gene is affected by vitamin A. The encoded protein of this gene may be associated with the cytoskeleton. A similar protein in rats may play a role in the regulation of cell differentiation. The rat protein binds and inhibits the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport. [provided by RefSeq]
Sequence: MDVNIAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVVAAMMISIVGFLSPFSpecifications
NP_006398 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.78kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DERP11/GTRAP3-18/HSPC127/JWA/PRAF3/addicsin/hp22/jmx | |
ARL6IP5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10550 | |
ARL6IP5 (Human) Recombinant Protein (Q01) | |
MDVNIAPLRAWDDFFPGSDRFARPDFRDISKWNNRVVSNLLYYQTNYLVVAAMMISIVGFLSPF | |
RUO | |
ARL6IP5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |