missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARHGEF12 Partial ORF (NP_056128, 817 a.a. - 916 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_056128 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23365 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124596
|
Abnova™
H00023365-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16114596
|
Abnova™
H00023365-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein is observed to form myeloid/lymphoid fusion partner in acute myeloid leukemia. [provided by RefSeq]
Sequence: VSREGILSPSELRKIFSNLEDILQLHIGLNEQMKAVRKRNETSVIDQIGEDLLTWFSGPGEEKLKHAAATFCSNQPFALEMIKSRQKKDSRFQTFVQDAESpecifications
NP_056128 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686O2372/KIAA0382/LARG/PRO2792 | |
ARHGEF12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23365 | |
ARHGEF12 (Human) Recombinant Protein (Q01) | |
VSREGILSPSELRKIFSNLEDILQLHIGLNEQMKAVRKRNETSVIDQIGEDLLTWFSGPGEEKLKHAAATFCSNQPFALEMIKSRQKKDSRFQTFVQDAE | |
RUO | |
ARHGEF12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |