missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APOA5 Full-length ORF (AAI01790.1, 1 a.a. - 366 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAI01790.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 116519 |
Molecular Weight (g/mol) | 67.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16127353
|
Abnova™
H00116519-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16137353
|
Abnova™
H00116519-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is an apolipoprotein and an important determinant of plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. This gene uses alternate polyadenylation sites and is located proximal to the apolipoprotein gene cluster on chromosome 11q23. [provided by RefSeq]
Sequence: MASMAAVLTWALALLSAFSATQARKGFWDYFSQTSGDKGRVEQIHQQKMAREPATLKDSLEQDLNNMNKFLEKLRPLSGSEAPRLPQDPVGMRRQLQEELEEVKARLQPYMAEAHELVGWNLEGLRQQLKPYTMDLMEQVALRVQELQEQLRVVGEDTKAQLLGGVDEAWALLQGLQSRVVHHTGRFKELFHPYAESLVSGIGRHVQELHRSVAPHAPASPARLSRCVQVLSRKLTLKAKALHARIQQNLDQLREELSRAFAGTGTEEGAGPDPQMLSEEVRQRLQAFRQDTYLQIAAFTRAIDQETEEVQQQLAPPPPGHSAFAPEFQQTDSGKVLSKLQARLDDLWEDITHSLHDQGHSHLGDPSpecifications
AAI01790.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APOA-V/APOAV/MGC126836/MGC126838/RAP3 | |
APOA5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
116519 | |
APOA5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
APOA5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |