missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human APCDD1 Partial ORF (NP_694545, 29 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_694545 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 147495 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102067
|
Abnova™
H00147495-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16192057
|
Abnova™
H00147495-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Sequence: HPDSRSHPRSLEKSAWRAFKESQCHHMLKHLHNGARITVQMPPTIEGHWVSTGCEVRSGPEFITRSYRFYHNNTFKAYQFYYGSNRCTNPTYTLIIRGKISpecifications
NP_694545 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B7323/DRAPC1/FP7019 | |
APCDD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
147495 | |
APCDD1 (Human) Recombinant Protein (Q01) | |
HPDSRSHPRSLEKSAWRAFKESQCHHMLKHLHNGARITVQMPPTIEGHWVSTGCEVRSGPEFITRSYRFYHNNTFKAYQFYYGSNRCTNPTYTLIIRGKI | |
RUO | |
APCDD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |