missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human AP4E1 Partial ORF (NP_031373, 1038 a.a. - 1137 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_031373
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 23431
Molecular Weight (g/mol) 36.74kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16164676
View Documents Promotion Details
Abnova™
H00023431-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16154676
View Documents Promotion Details
Abnova™
H00023431-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

The heterotetrameric adaptor protein (AP) complexes sort integral membrane proteins at various stages of the endocytic and secretory pathways. AP4 is composed of 2 large chains, beta-4 (AP4B1; MIM 607245) and epsilon-4 (AP4E1), a medium chain, mu-4 (AP4M1; MIM 602296), and a small chain, sigma-4 (AP4S1; MIM 607243).[supplied by OMIM]

Sequence: SDDFGKLWLSFANDVKQNVKMSESQAALPSALKTLQQKLRLHIIEIIGNEGLLACQLLPSIPCLLHCRVHADVLALWFRSSCSTLPDYLLYQCQKVMEGS
Specifications

Specifications

NP_031373
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686L12167
AP4E1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
23431
AP4E1 (Human) Recombinant Protein (Q01)
SDDFGKLWLSFANDVKQNVKMSESQAALPSALKTLQQKLRLHIIEIIGNEGLLACQLLPSIPCLLHCRVHADVLALWFRSSCSTLPDYLLYQCQKVMEGS
RUO
AP4E1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications