missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ANXA9 Partial ORF (NP_003559.1, 270 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008416-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16128515

  • 545.00€ / 25µg

Estimated Shipment: 16-09-2022
to see stock.
Add to basket

Description

Description

The annexins are a family of calcium-dependent phospholipid-binding proteins. Members of the annexin family contain 4 internal repeat domains, each of which includes a type II calcium-binding site. The calcium-binding sites are required for annexins to aggregate and cooperatively bind anionic phospholipids and extracellular matrix proteins. This gene encodes a divergent member of the annexin protein family in which all four homologous type II calcium-binding sites in the conserved tetrad core contain amino acid substitutions that ablate their function. However, structural analysis suggests that the conserved putative ion channel formed by the tetrad core is intact. [provided by RefSeq]

Sequence: DKLHQALQETEPNYQVLIRILISRCETDLLSIRAEFRKKFGKSLYSSLQDAVKGDCQSALLALCRAEDM
Specifications

Specifications

NP_003559.1
Liquid
8416
ANXA9 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ANX31
ANXA9
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.33kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DKLHQALQETEPNYQVLIRILISRCETDLLSIRAEFRKKFGKSLYSSLQDAVKGDCQSALLALCRAEDM
RUO
ANXA9
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers