Learn More
Abnova™ Human AMOT Partial ORF (NP_573572, 361 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_573572 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 154796 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16102207
|
Abnova™
H00154796-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16192197
|
Abnova™
H00154796-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
This gene belongs to the motin family of angiostatin binding proteins characterized by conserved coiled-coil domains and C-terminal PDZ binding motifs. The encoded protein is expressed predominantly in endothelial cells of capillaries as well as larger vessels of the placenta where it may mediate the inhibitory effect of angiostatin on tube formation and the migration of endothelial cells toward growth factors during the formation of new blood vessels. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: RKEPSKTEQLSCMRPAKSLMSISNAGSGLLSHSSTLTGSPIMEEKRDDKSWKGSLGILLGGDYRAEYVPSTPSPVPPSTPLLSAHSKTGSRDCSTQTERGSpecifications
NP_573572 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1071 | |
AMOT | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
154796 | |
AMOT (Human) Recombinant Protein (Q01) | |
RKEPSKTEQLSCMRPAKSLMSISNAGSGLLSHSSTLTGSPIMEEKRDDKSWKGSLGILLGGDYRAEYVPSTPSPVPPSTPLLSAHSKTGSRDCSTQTERG | |
RUO | |
AMOT | |
Wheat Germ (in vitro) | |
GST | |
Liquid |