missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AMFR Partial ORF (NP_001135, 451 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001135 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 267 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111541
|
Abnova™
H00000267-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
16101541
|
Abnova™
H00000267-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
Autocrine motility factor is a tumor motility-stimulating protein secreted by tumor cells. The protein encoded by this gene is a glycosylated transmembrane protein and a receptor for autocrine motility factor. The receptor, which shows some sequence similarity to tumor protein p53, is localized to the leading and trailing edges of carcinoma cells. [provided by RefSeq]
Sequence: QASNSQLNAMAHQIQEMFPQVPYHLVLQDLQLTRSVEITTDNILEGRIQVPFPTQRSDSIRPALNSPVERPSSDQEEGETSAQTERVPLDLSPRLEETLDSpecifications
NP_001135 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GP78/RNF45 | |
AMFR | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
267 | |
AMFR (Human) Recombinant Protein (Q01) | |
QASNSQLNAMAHQIQEMFPQVPYHLVLQDLQLTRSVEITTDNILEGRIQVPFPTQRSDSIRPALNSPVERPSSDQEEGETSAQTERVPLDLSPRLEETLD | |
RUO | |
AMFR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |