Learn More
Abnova™ Human AKT3 Partial ORF (AAD29089, 100 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAD29089 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10000 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111606
|
Abnova™
H00010000-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16101606
|
Abnova™
H00010000-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a member of the AKT, also called PKB, serine/threonine protein kinase family. AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors. They are involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, as well as glycogen synthesis and glucose uptake. This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). Alternatively splice transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: EAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDESpecifications
AAD29089 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434N0250/PKB-GAMMA/PKBG/PRKBG/RAC-PK-gamma/RAC-gamma/STK-2 | |
AKT3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
10000 | |
AKT3 (Human) Recombinant Protein (Q01) | |
EAIQAVADRLQRQEEERMNCSPTSQIDNIGEEEMDASTTHHKRKTMNDFDYLKLLGKGTFGKVILVREKASGKYYAMKILKKEVIIAKDE | |
RUO | |
AKT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |