missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AGPAT1 Full-length ORF (NP_006402.1, 1 a.a. - 283 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_006402.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10554 |
Molecular Weight (g/mol) | 58.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135322
|
Abnova™
H00010554-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16125322
|
Abnova™
H00010554-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
This gene encodes an enzyme that converts lysophosphatidic acid (LPA) into phosphatidic acid (PA). LPA and PA are two phospholipids involved in signal transduction and in lipid biosynthesis in cells. This enzyme localizes to the endoplasmic reticulum. This gene is located in the class III region of the human major histocompatibility complex. Alternative splicing results in two transcript variants encoding the same protein. [provided by RefSeq]
Sequence: MDLWPGAWMLLLLLFLLLLFLLPTLWFCSPSAKYFFKMAFYNGWILFLAVLAIPVCAVRGRNVENMKILRLMLLHIKYLYGIRVEVRGAHHFPPSQPYVVVSNHQSSLDLLGMMEVLPGRCVPIAKRELLWAGSAGLACWLAGVIFIDRKRTGDAISVMSEVAQTLLTQDVRVWVFPEGTRNHNGSMLPFKRGAFHLAVQAQVPIVPIVMSSYQDFYCKKERRFTSGQCQVRVLPPVPTEGLTPDDVPALADRVRHSMLTVFREISTDGRGGGDYLKKPGGGGSpecifications
NP_006402.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
1-AGPAT1/G15/LPAAT-alpha/LPAATA/MGC4007/MGC5423 | |
AGPAT1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10554 | |
AGPAT1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AGPAT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |