missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AGA Full-length ORF (NP_000018.1, 1 a.a. - 346 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_000018.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 175 |
Molecular Weight (g/mol) | 63.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16111351
|
Abnova™
H00000175-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16101351
|
Abnova™
H00000175-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
Aspartylglucosaminidase is involved in the catabolism of N-linked oligosaccharides of glycoproteins. It cleaves asparagine from N-acetylglucosamines as one of the final steps in the lysosomal breakdown of glycoproteins. The lysosomal storage disease aspartylglycosaminuria is caused by a deficiency in the AGA enzyme. [provided by RefSeq]
Sequence: MARKSNLPVLLVPFLLCQALVRCSSPLPLVVNTWPFKNATEAAWRALASGGSALDAVESGCAMCEREQCDGSVGFGGSPDELGETTLDAMIMDGTTMDVGAVGDLRRIKNAIGVARKVLEHTTHTLLVGESATTFAQSMGFINEDLSTSASQALHSDWLARNCQPNYWRNVIPDPSKYCGPYKPPGILKQDIPIHKETEDDRGHDTIGMVVIHKTGHIAAGTSTNGIKFKIHGRVGDSPIPGAGAYADDTAGAAAATGNGDILMRFLPSYQAVEYMRRGEDPTIACQKVISRIQKHFPEFFGAVICANVTGSYGAACNKLSTFTQFSFMVYNSEKNQPTEEKVDCISpecifications
NP_000018.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGU/ASRG/GA | |
AGA | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
175 | |
AGA (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
AGA | |
Wheat Germ (in vitro) | |
GST | |
Liquid |