missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADMR Partial ORF (NP_009195, 1 a.a. - 54 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_009195 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11318 |
Molecular Weight (g/mol) | 31.68kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116532
|
Abnova™
H00011318-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16106532
|
Abnova™
H00011318-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Adrenomedullin is a potent vasodilator peptide that exerts major effects on cardiovascular function. This gene encodes a seven-transmembrane protein that belongs to the family 1 of G-protein coupled receptors. Studies of the rat counterpart suggest that the encoded protein may function as a receptor for adrenomedullin. [provided by RefSeq]
Sequence: MSVKPSWGPGPSEGVTAVPTSDLGEIHNWTELLDLFNHTLSECHVELSQSTKRVSpecifications
NP_009195 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.68kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
7TMR/ADMR/AM-R/AMR/G10D/MGC34399/gamrh/hrhAMR | |
GPR182 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11318 | |
ADMR (Human) Recombinant Protein (Q01) | |
MSVKPSWGPGPSEGVTAVPTSDLGEIHNWTELLDLFNHTLSECHVELSQSTKRV | |
RUO | |
GPR182 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |