missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ADCY8 Partial ORF (NP_001106, 1142 a.a. - 1251 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_001106
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 114
Molecular Weight (g/mol) 37.84kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16114174
View Documents Promotion Details
Abnova™
H00000114-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16124174
View Documents Promotion Details
Abnova™
H00000114-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. The enzymatic activity is under the control of several hormones, and different polypeptides participate in the transduction of the signal from the receptor to the catalytic moiety. Stimulatory or inhibitory receptors (Rs and Ri) interact with G proteins (Gs and Gi) that exhibit GTPase activity and they modulate the activity of the catalytic subunit of the adenylyl cyclase [provided by RefSeq]

Sequence: DQGFAFDYRGEIYVKGISEQEGKIKTYFLLGRVQPNPFILPPRRLPGQYSLAAVVLGLVQSLNRQRQKQLLNENNNTGIIKGHYNRRTLLSPSGTEPGAQAEGTDKSDLP
Specifications

Specifications

NP_001106
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AC8/ADCY3/HBAC1
ADCY8
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
114
ADCY8 (Human) Recombinant Protein (Q01)
DQGFAFDYRGEIYVKGISEQEGKIKTYFLLGRVQPNPFILPPRRLPGQYSLAAVVLGLVQSLNRQRQKQLLNENNNTGIIKGHYNRRTLLSPSGTEPGAQAEGTDKSDLP
RUO
ADCY8
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications