Learn More
Abnova™ Human ADAMTS15 Partial ORF (NP_620686, 863 a.a. - 950 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_620686 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 170689 |
Molecular Weight (g/mol) | 35.42kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132327
|
Abnova™
H00170689-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16122327
|
Abnova™
H00170689-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the proteins encoded by ADAMTS1 and ADAMTS8. [provided by RefSeq]
Sequence: AVDCRGSAGQRTVPACDAAHRPVETQACGEPCPTWELSAWSPCSKSCGRGFQRRSLKCVGHGGRLLARDQCNLHRKPQELDFCVLRPCSpecifications
NP_620686 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126403 | |
ADAMTS15 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
170689 | |
ADAMTS15 (Human) Recombinant Protein (Q01) | |
AVDCRGSAGQRTVPACDAAHRPVETQACGEPCPTWELSAWSPCSKSCGRGFQRRSLKCVGHGGRLLARDQCNLHRKPQELDFCVLRPC | |
RUO | |
ADAMTS15 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |