missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ADAM30 Partial ORF (NP_068566, 199 a.a. - 298 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_068566 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 11085 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16193676
|
Abnova™
H00011085-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16103686
|
Abnova™
H00011085-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats. [provided by RefSeq]
Sequence: YKHPKYLELILLFDQSRYRFVNNNLSQVIHDAILLTGIMDTYFQDVRMRIHLKALEVWTDFNKIRVGYPELAEVLGRFVIYKKSVLNARLSSDWAHLYLQSpecifications
NP_068566 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
svph4 | |
ADAM30 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
11085 | |
ADAM30 (Human) Recombinant Protein (Q01) | |
YKHPKYLELILLFDQSRYRFVNNNLSQVIHDAILLTGIMDTYFQDVRMRIHLKALEVWTDFNKIRVGYPELAEVLGRFVIYKKSVLNARLSSDWAHLYLQ | |
RUO | |
ADAM30 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |