missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ACTR1B Partial ORF (NP_005726.1, 191 a.a. - 285 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_005726.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10120
Molecular Weight (g/mol) 36.08kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16161906
View Documents Promotion Details
Abnova™
H00010120-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16151906
View Documents Promotion Details
Abnova™
H00010120-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene encodes a 42.3 kD subunit of dynactin, a macromolecular complex consisting of 10 subunits ranging in size from 22 to 150 kD. Dynactin binds to both microtubules and cytoplasmic dynein and is involved in a diverse array of cellular functions, including ER-to-Golgi transport, the centripetal movement of lysosomes and endosomes, spindle formation, chromosome movement, nuclear positioning, and axonogenesis. This subunit, like ACTR1A, is an actin-related protein. These two proteins, which are of equal length and share 90% amino acid identity, are present in a constant ratio of approximately 1:15 in the dynactin complex. [provided by RefSeq]

Sequence: SRYLRLLLRKEGVDFHTSAEFEVVRTIKERACYLSINPQKDEALETEKVQYTLPDGSTLDVGPARFRAPELLFQPDLVGDESEGLHEVVAFAIHK
Specifications

Specifications

NP_005726.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARP1B/CTRN2/PC3
ACTR1B
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
10120
ACTR1B (Human) Recombinant Protein (Q01)
SRYLRLLLRKEGVDFHTSAEFEVVRTIKERACYLSINPQKDEALETEKVQYTLPDGSTLDVGPARFRAPELLFQPDLVGDESEGLHEVVAFAIHK
RUO
ACTR1B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications