Learn More
Abnova™ Human ACTA2 Full-length ORF (AAH17554.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH17554.1 |
---|---|
For Use With (Application) | Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 59 |
Molecular Weight (g/mol) | 67.21kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16191201
|
Abnova™
H00000059-P01.10UG |
10μg |
335.00€
10µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16101211
|
Abnova™
H00000059-P01.25UG |
25μg |
508.00€
25µg |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Defects in this gene cause aortic aneurysm familial thoracic type 6. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]
Sequence: MCEEEDSTALVCDNGSGLCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIITNWDDMEKIWHHSFYNELRVAPEEHPTLLTEAPLNPKANREKMTQIMFETFNVPAMYVAIQAVLSLYASGRTTGIVLDSGDGVTHNVPIYEGYALPHAIMRLDLAGRDLTDYLMKILTERGYSFVTTAEREIVRDIKEKLCYVALDFENEMATAASSSSLEKSYELPDGQVITIGNERFRCPETLFQPSFIGMESAGIHETTYNSIMKCDIDIRKDLYANNVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKQEYDEAGPSIVHRKCFSpecifications
AAH17554.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67.21kDa | |
Glutathione Sepharose 4 Fast Flow | |
Store at −80°C. Aliquot to avoid repeated freezing and thawing. | |
AAT6, ACTSA | |
ACTA2 | |
Recombinant | |
wheat germ expression system |
Protein Array, ELISA, Western Blot | |
59 | |
Human ACTA2 Full-length ORF (AAH17554.1, 1 a.a. - 377 a.a.) Recombinant Protein with GST-tag at N-terminal | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RUO | |
ACTA2 | |
Wheat Germ (in vitro) | |
GST Tag | |
Liquid |