missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HSD11B1 (Human) Recombinant Protein
Human HSD11B1 full-length ORF ( AAH12593, 1 a.a. - 292 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003290-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAFMKKYLLPILGLFMAYYYYSANEEFRPEMLQGKKVIVTGASKGIGREMAYHLAKMGAHVVVTARSKETLQKVVSHCLELGAASAHYIAGTMEDMTFAEQFVAQAGKLMGGLDMLILNHITNTSLNLFHDDIHHVRKSMEVNFLSYVVLTVAALPMLKQSNGSIVVVSSLAGKVAYPMVAAYSASKFALDGFFSSIRKEYSVSRVNVSITLCVLGLIDTETAMKAVSGIVHMQAAPKEECALEIIKGGALRQEEVYYDSSLWTTLLIRNPCRKILEFLYSTSYNMDRFINK
Specifications
AAH12593 | |
hydroxysteroid (11-beta) dehydrogenase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSD11B1 | |
GST |
3290 | |
Wheat germ expression system | |
25 μg | |
11-DH, 11-beta-HSD1, HDL, HSD11, HSD11B, HSD11L, MGC13539, SDR26C1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |