missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His Tag, Armenian Hamster anti-Non-species specific, Clone: 3D5, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals NBP2-81022-0.2MG
This item is not returnable.
View return policy
Description
His Tag Monoclonal specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.Specifications
His Tag | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
Purified his tagged Fab fragment. | |
0.2 mg | |
Primary | |
Non-species specific | |
Purified |
Western Blot, ELISA | |
3D5 | |
Unconjugated | |
PBS | |
Armenian Hamster | |
Protein A purified | |
RUO | |
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |