Learn More
Honeywell™ HIKE (i)XTREM S3 Shoes
Honeywell™ HIKE (i)XTREM S3 Shoes are constructed of water-resistant full grain leather.
Brand:  Honeywell™ 6246201-42/7
Additional Details : Weight : 1.62000kg
 View more versions of this product
Product Code. 15816565
  • 118.00€ / 1 pair
Estimated Shipment: 11-10-2021
Add to basket
Description
Description
  • Water resistant grain full leather
  • Ergonomic collar and padded tongue for an optimized comfort
  • Gusset to avoid water and dust penetration
  • Breathable 3D lining
  • Overcap
  • Toe cap metal free
  • Metal free antiperforation insert
  • Injected PU / Nitrile sole
  • (i)NTELLIGENT wear control
  • EN ISO 20345:2011 S3 HRO HI CI SRC
Specifications
Specifications
EN ISO 20345:2011 S3 HRO HI CI SRC
Leather
Polyurethane and Nitrile
Brown, Black
42
Metal-free