Learn More
Abeba™ High Original 7009 Shoes
Animal fats, oil and fuel resistant. Sanitized™ treated.
Brand:  Abeba™ 7009/38
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 15622468
  • 35.45€ / 1 pair
Estimated Shipment
03-07-2020
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E, FO SRC
  • Smooth leather, perforated
  • Black
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather top layer
  • Narrow fit
  • Wedge heel 45 mm
Specifications
Specifications
Animal fats, oil and fuel
Black
38
Polyurethane
CE EN ISO 20347:2012 OB A, E, FO SRC
Leather
Slip-resistant
7009