Specifications
| Product Type | Stackable Pail |
| Material | HDPE |
| Certifications/Compliance | FDA Certified |
| Height (Metric) | 368.3 mm |
| Height (Metric) With Closure | 368.3 mm |
| Quantity | 1 Ea. |
| Width (Metric) | 247.6 mm |