missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-2Kb Mouse anti-Mouse, PE, Clone: AF6-88.5, BD
Mouse Monoclonal Antibody
151.00€ - 397.00€
Specifications
Antigen | H-2Kb |
---|---|
Clone | AF6-88.5 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15800749
|
BD Biosciences
561072 |
25 μg |
151.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
15807288
|
BD Biosciences
553570 |
0.1 mg |
397.00€
Each |
Estimated Shipment: 22-04-2024
Log in to see stock. |
|||||
Description
The AF6-88.5 antibody reacts with the H-2Kb MHC class I alloantigen. Reactivity with other haplotypes (e.g., d, f, j, k, p, q, r, s, u, v) has not been observed.
Specifications
H-2Kb | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
Mouse C57BL splenocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
AF6-88.5 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.