Learn More
GSTM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557191
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: TYDILDQNRI FDPKCLDEFP NLKAFMCRFE ALEKIAAYLQ SDQFCKMPIN N Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.
GSTM3 Polyclonal antibody specifically detects GSTM3 in Human samples. It is validated for Immunohistochemistry.Specifications
GSTM3 | |
Polyclonal | |
Unconjugated | |
GSTM3 | |
brain GST; brain type mu-glutathione S-transferase; Fsc2; glutathione S-alkyltransferase M3; glutathione S-aralkyltransferase M3; glutathione S-aryltransferase M3; glutathione S-transferase GT9.3; glutathione S-transferase M3 (brain); glutathione S-transferase Mu 3; glutathione S-transferase mu 3 (brain); glutathione S-transferase, mu 3; glutathione S-transferase, Mu-3; GST class-mu 3; GST5; GSTB; GSTM3; GSTM3-3; GTM3; hGSTM3-3; mGSTM5; S-(hydroxyalkyl)glutathione lyase M3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P21266 | |
GSTM3 | |
Recombinant protein corresponding to Human GSTM3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only