Learn More
Sartorius™ Grade 1289 Wet-strengthened Qualitative Filter Papers
Used for analytical purposes and routine analyses, whenever no gravimetric analyses are required
Brand:  Sartorius™ FT-2-207-580580
Additional Details : Weight : 0.10000kg
Product Code. 10042331
  • 340.00€ / Pack of 100
Estimated Shipment: 14-05-2024
Description
  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N <0.1%)
  • Wet-strengthened
  • Supplied in sheets
Specifications
Specifications
Sheet
8 to 12 μm
Used for analytical purposes and routine analyses, whenever no gravimetric analyses are required
100/Pk.
Qualitative Filter Paper
580 mm
Cotton Linters, Refined Pulp
20μm/sec.
1289
0.21 mm
580 L x 580 Wmm
580 mm