Learn More
Goat anti-Mouse IgG1, IgG2a, IgG2b, IgG3 Cross-Adsorbed Secondary Antibody, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen 31232
Additional Details : Weight : 0.00010kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31232 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31232 reacts with the heavy chains of mouse IgG subclasses 1, 2a, 2b and 3, but not with the light chains of most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with human, bovine, and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA
Mouse IgG1, IgG2a, IgG2b, IgG3 Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG1, IgG2a, IgG2b, IgG3 Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
Mouse IgG1, IgG2a, IgG2b, IgG3 Cross-Adsorbed | |
Polyclonal | |
Unconjugated | |
Goat | |
1 mg | |
Secondary | |
4° C | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.3 mg/mL | |
PBS with no preservative; pH 7.6 | |
Antigen affinity chromatography | |
RUO | |
Mouse | |
Liquid |