missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse Complement C3 Polyclonal Anitbody, MP Biomedicals
Brand: MP Biomedicals 0855463
Additional Details : Weight : 0.00500kg
Description
Host Species: Goat
Species Reactivity: Mouse
Specifications
Complement C3 | |
Unconjugated | |
5 mL | |
Mouse |
Polyclonal | |
Goat | |
Primary | |
Lyophilized |