missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Dog IgG, Whole Molecule Polyclonal Secondary Antibody, FITC, MP Biomedicals
Brand: MP Biomedicals 0855325
Additional Details : Weight : 0.00200kg
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Dog
Specifications
IgG, Whole Molecule | |
FITC | |
2 mL | |
Canine | |
IgG |
Polyclonal | |
Goat | |
Secondary | |
Purified |