Learn More
R&D Systems™ GMP N-2 MAX Media Supplement (100X), Animal-free
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Specifications
Specifications
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 5 mL |
| Species | Multi-Species |