Learn More
Sartorius Stedim Biotech™ Glass Microfiber Filters, Grade 1336/2
Generally used for air and gas monitoring, but are suitable as a prefilter.
Brand:  Sartorius Stedim Biotech™ FT-201120-210297
Additional Details : Weight : 0.45600kg
Product Code. 15922820
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Description
Description
  • Produced with synthetic binders to make them more resistant
  • Suitable for air monitoring or prefiltration
  • Non-sterile, bulk packed
Specifications
Specifications
Sheet
Non-sterile
Hydrophobic
0.4 mm
Glass Microfiber with Binder
1336/2
100/Pk.
210 x 297 mm