Learn More
Cytiva RNAspin Midi Isolation Kit
Yields high-quality total RNA due to removal of genomic DNA through on-column DNase I treatment
Brand: Cytiva 25-0500-73
Additional Details : CAS : 9048-46-8 Weight : 1.08800kg Transport : UN number : 1993 Chem class : 3 Pack group : III
Includes: Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I
Description
- Prefilters included to reduce lysate viscosity
- Scalable input and output
- Binding capacity: 700μg
- Sample size (animal tissue): 200mg
- Sample size (cell culture): <5 × 107 cells
- Elution volume: 500μL
- RNA Purity (A260/A280): 1.8 to 2.2
- Preparation time: 80 min./4 preps
- For in vitro use only
Sensitive downstream applications, isolation of total RNA from cultured cells and tissue
Specifications
illustra RNAspin Midi RNA Isolation Kit is designed for for the rapid extraction and purification of RNA from a wide range of starting material. | |
Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I, prefilters | |
illustra RNAspin Midi Kit | |
<200 mg | |
Up to 700μg |
Suitable for use in sensitive downstream applicationsSuitable for in vitro use onlyRecommended for the isolation of total RNA from cultured cells and tissue | |
20 Preps | |
1.8-2.2 | |
80 min/4 preps |
Safety and Handling
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .