Learn More
Memmert™ Gas Connections
Memmert™ Gas Connections with quick release connectors for automatic switch-over of gas cylinders.
Brand:  Memmert™ ICO150T1
Additional Details : Weight : 2.80000kg
 View more versions of this product
Product Code. 15622224
  • 320.00€ / Each
please call +351214253350
Add to basket
Description
Description
  • CO2 connections set includes a hose with coupling and clamp
  • Quick release
Specifications
Specifications
ICO150
Gas Connections
Two CO2 connections set
Videos