Learn More
Fungilab™ Stainless Steel Spindles
Accurately measure viscosities with these stainless steel spindles. Stainless Steel Spindles come in a variety of volumes and viscosity ranges.
Brand:  Fungilab™ V12400N
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11876471
Quantity Price
1 498.43€ / Each
6 438.72€ / Each
Estimated Shipment
27-01-2021
Add to basket
Description
Description
Spindles for use with Small Sample Adapters (Cat. Nos. 06-815-032, -031).
  • This system is used for measurements in accordance with the ASTM-ISO 2555 standard
  • Standard spindles (included in standard delivery): R and H spindles, L spindles
  • Special spindles (additionally ordered with small sample adapters APM and APM/B): TL and TR spindles
  • Spindle set L: 20 to 6,000,000 cP (for ADV and PRO models); 20 to 2,000,000 cP (for ONE model)
  • Spindle set R: 100 to 40,000,000cP
  • Spindles set R for H models: 200 to 106,000,000cP
Specifications
Specifications
ASTM, ISO 2555
Stainless Steel
Small sample adapter
For relative viscosity measurement
3 to 60rpm
L1, L2, L3, L4 Spindle