Learn More
Fungilab™ Glass Thermometers for Viscoball and Hoppler Viscometer
Complete a Viscoball or Hoppler Viscometer setup with the appropriate thermometer of your choice. Glass Thermometers for Viscoball and Hoppler Viscometer come in a variety of temperature ranges.
Brand:  Fungilab™ V91003
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11856591
Quantity Price
1 123.55€ / Each
10 116.12€ / Each
Estimated Shipment
18-01-2021
Add to basket
Description
Description
  • Can be used with Fungilab Visco Ball Viscometer
  • Divisions: 0.1°C
Specifications
Specifications
DIN 53015, ISO 12058
49° to 76°C
Thermometer
Glass
Hoppler viscometer