Learn More
Fungilab™ ASTM D-1200 Flow Cup Viscometers
Measure the viscosity of paint, ink, varnishes, and similar products industries. ASTM D-1200 Flow Cup Viscometers are available in a variety of sizes.
Brand:  Fungilab™ FCA004
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11846441
Quantity Price
1 272.45€ / Each
6 247.11€ / Each
Estimated Shipment
18-01-2021
Add to basket
Description
Description
  • Provides kinematic viscosity data
  • Data is generally expressed in seconds flow time when the fluid is flowing through an oriface at a specified temperature
  • The viscosity is calculated from flow time to centistrokes (cSt)
  • The calibration certificates are supplied with the cups
Specifications
Specifications
4
ASTM D1200, calibration certificate
Anodized aluminum body, stainless steel orifice
Used in paint, ink, varnishes and similar products industries
70 to 370 cSt
4.12mm
Calibration certificate
4
Flow Cup Viscometer