missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Honeywell Fluka™

Catalog Number 15622830
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 x 2.5 L
70 kg
1200 kg
This item is not returnable. View return policy
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O
This item is not returnable. View return policy
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
formic acid
C(=O)O

Puriss., meets analytical specifications of DAC, FCC, 98.0-100%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Linear Formula HCOOH
UN Number 1779
Beilstein 1209246
Formula Weight 46.03g/mol
Percent Purity 98 to 100%
Grade puriss.
Density 1.22g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.57
Flash Point 48°C (118.4°F)
Ignition Point 1004°F
Vapor Density 1.6 (vs air)
Vapor Pressure 44.8mmHg (20°C)
Chemical Name or Material Formic Acid, Meets analytical specifications of DAC, FCC
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'