Learn More
Flavin Adenine Dinucleotide Disodium Salt Hydrate, ≥95% (dry basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Plastic bottle
Specifications
Physical Form | Crystalline Powder |
Quantity | 500 mg |
IUPAC Name | disodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[(2R,3S,4S)-5-{7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl}-2,3,4-trihydroxypentyl phosphonato]oxy}phosphonate |
Formula Weight | 829.51 (anhydrous) |
Percent Purity | ≥93% |
Packaging | Plastic bottle |
Assay | Dry basis |
Chemical Name or Material | Flavin Adenine Dinucleotide Disodium Salt Hydrate |